KIMBERLY-CLARK® Universal Precautions Lab Coats, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA10815-732
Supplier: Kimberly-Clark
Specifications
- Clothing Size:M
- Garment application:Laboratory
- Color:White
- Cat. no.:CA10815-732
- Supplier No.:10041
Specifications
About this item
Disposable, fluid-resistant lab coats are manufactured from a protective three-layer spunbond-meltblown-spunbond (SMS) fabric that offers exceptional resistance to blood strikethrough for moderate fluid contact
- Disposable, fluid-resistant, three-layer SMS fabric
- Six-snap front closure, three reinforced pockets, back vent, traditional collar, and knit cuffs
- Tailored styling and fit enhances worker comfort
- Non sterile, latex-free
Lab coats feature a six-snap front closure, three reinforced pockets, back vent, traditional collar, and knit cuffs. Tailored styling and fit enhances worker comfort. Nonsterile. Latex-free.
Customers Who Bought This Also Bought