Order Entry
ContactUsLinkComponent
Perfecta QPCR Fastmix LRX 1250
PerfeCTa® qPCR FastMix® UNG Reaction Mixes, Quantabio
Catalog #: CA101419-220
Supplier:  Quantabio
CAS Number:  
PerfeCTa® qPCR FastMix® UNG Reaction Mixes, Quantabio
Catalog #: CA101419-220
Supplier:  Quantabio
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
    qPCR probe
  • Description:
    PerfeCTa® qPCR FastMix®, Low ROX™
  • Cat. no.:
    CA101419-220
  • No. of Reactions:
    1250
  • Supplier No.:
    95078-012

Specifications

About this item

FastMix® reaction mixes include buffers and stabilizers optimized for maximum PCR efficiency, reproducibility, and sensitivity with TaqMan® and TaqMan® MGB probe chemistries

  • Fast Cycling protocols
  • Contains UNG to eliminate amplification of carry-over contamination

Kits are available for all real-time PCR instrument platforms including those requiring normalization with ROX™ reference dye. Kits 101419-202, -204, and -206 are for use with platforms not requiring passive reference dye, including Roche LightCycler® 480, Bio-Rad® iQ™, MyiQ™, iQ™5, Opticon®, and Chromo4™, Corbett Rotor-Gene™, Eppendorf® Mastercycler®, and Cepheid® SmartCycler®. Kits with Low ROX™ reference dye are for use with platforms including AB 7500 and Stratagene® Mx™. Kits with ROX™ reference dye are for use with platforms including AB 7000, 7300, 7700, 7900, and StepOne™.

A key component of PerfeCTa® qPCR FastMix® reaction mixes is AccuFast™ Taq DNA polymerase that enables instant activation and rapid polymerization kinetics. Upon heat activation (20 seconds at 95°C), the antibodies irreversibly denature releasing fully active, unmodified Taq DNA polymerase, which promotes specific and efficient primer extension with the convenience of room temperature reaction assembly. FastMix® reaction mixes deliver maximum PCR efficiency, sensitivity, specificity, and a robust fluorescent signal with either fast or conventional cycling conditions.

Customers Who Bought This Also Bought