Order Entry
ContactUsLinkComponent
DNA/RNA Isolation Kit
E.Z.N.A. DNA/RNA Isolation Kit, Omega Bio-tek
Catalog #: CA101414-838
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A. DNA/RNA Isolation Kit, Omega Bio-tek
Catalog #: CA101414-838
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    Copurification of DNA & RNA
    DNA
    RNA
  • Description:
    E.Z.N.A. DNA/RNA Isolation Kit
  • Size:
    50 Preps
  • Description:
    E.Z.N.A. DNA/RNA Isolation Kit
  • Cat. no.:
    CA101414-838
  • Supplier No.:
    R6731-01

Specifications

About this item

The E.Z.N.A DNA/RNA Isolation Kit is designed for the simultaneous isolation of both genomic DNA and total RNA from the same cells or tissues. The sample is first lysed and homogenized in a special denaturing buffer, spun to pellet RNA and undigested particles, and then the supernatant is applied to a HiBind DNA spin column to bind DNA. The RNA pellet is dissolved and purified by a HiBind RNA spin column. Since there is no need to divide the sample into two parts for separate purification procedures, maximum yield of DNA and RNA can be purified from the entire sample.

Customers Who Bought This Also Bought