Order Entry
ContactUsLinkComponent
PerfeCTa Sybr Fastmx Lrx 5000
PerfeCTa® SYBR® Green FastMix® Reaction Mixes, Quantabio
Catalog #: CA101414-292
Supplier:  Quantabio
CAS Number:  
PerfeCTa® SYBR® Green FastMix® Reaction Mixes, Quantabio
Catalog #: CA101414-292
Supplier:  Quantabio
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • 03_Application:
    qPCR dye
  • Description:
    PerfeCTa® SYBR® Green FastMix®, Low ROX™
  • Cat. no.:
    CA101414-292
  • No. of Reactions:
    5000
  • Supplier No.:
    95074-05K

Specifications

About this item

This unique combination of proprietary buffer, stabilizers, and AccuFast™ Taq DNA polymerase delivers maximum PCR efficiency, sensitivity, and specificity, along with a robust fluorescent signal using fast, or conventional, cycling protocols with SYBR® Green qPCR

  • 2× concentrated reagents minimize pipetting steps and improve accuracy
  • Ultrapure, hot start AccuStart enzyme technology and anti-foaming technology
  • FastMix formulation supports both fast and standard thermal cycling conditions

AccuFast™ Taq DNA polymerase is a key component of this reaction mix. This hot-start Taq contains a unique mixture of monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step (>48 hours at room temperature). Similar to our AccuStart™ Taq DNA polymerase, these antibodies are irreversibly inactivated during the initial PCR denaturation step. However, unlike other antibody hot-start polymerases, activation of AccuFast™ Taq is instantaneous at 95°C. Rapid recovery of fully active, unmodified Taq DNA polymerase is critical for efficient extension kinetics. Replication of fragments up to 200bp is complete in less than 20 seconds at 60°C.

Kits are available for all real-time PCR instrument platforms including those requiring normalization with ROX™ reference dye. Platforms not requiring passive reference dye include Roche LightCycler® 480, Bio-Rad® iQ™, MyiQ™, iQ™5, Opticon®, and Chromo4™, Corbett Rotor-Gene®, Eppendorf® Mastercycler®, and Cepheid® SmartCycler®. Kits with Low ROX™ reference dye are for use with platforms including AB 7500 and Stratagene® Mx™. Kits with ROX™ reference dye are for use with platforms including AB 7000, 7300, 7700, 7900, and StepOne™.

Highly specific amplification is crucial to successful qPCR with SYBR® Green I dye technology because this dye binds to and detects any dsDNA generated during amplification.

Customers Who Bought This Also Bought