Order Entry
ContactUsLinkComponent
Qscript 1-Step QRT-PCR, 200R
qScript™ One-Step qRT-PCR Kits
Catalog #: CA101414-172
Supplier:  Quantabio
CAS Number:  
Qscript 1-Step QRT-PCR, 200R
qScript™ One-Step qRT-PCR Kits
Catalog #: CA101414-172
Supplier:  Quantabio
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
    RT-qPCR probe
  • One step:
    Yes
  • Description:
    qScript™ One-Step qRT-PCR
  • Cat. no.:
    CA101414-172
  • No. of Reactions:
    200
  • Supplier No.:
    95057-200

Specifications

About this item

Synthesis of cDNA and PCR amplification are carried out in the same tube without opening between procedures

  • One-step convenience
  • High efficiency and sensitivity
  • Available with reference dye pre-mixed

Kits are available for all real-time PCR instrument platforms including those requiring normalization with ROX™ reference dye. Kits 101414-170 and -172 are for use with platforms not requiring passive reference dye, including Roche LightCycler® 480, Bio-Rad® iQ™, MyiQ™, iQ™5, Opticon®, Chromo4™, Corbett Rotor-Gene®, Eppendorf® Mastercycler®, and Cepheid® SmartCycler®. Kits with ROX™ reference dye are for use with platforms including AB 7000, 7300, 7700, and 7900 and StepOne™. Kits with Low ROX™ reference dye are for use with platforms including AB 7500 and Stratagene® Mx™.

Systems have been optimized to deliver maximum RT-PCR efficiency, sensitivity, and specificity, enabling unbiased co-amplification of low copy transcripts in the presence of higher copy reference genes. The proprietary reaction buffer has been specifically formulated to maximize activities of both reverse transcriptase and Taq DNA polymerase while minimizing the potential for primer-dimer and other non-specific PCR artifacts. Highly specific amplification is crucial to successful qRT-PCR as non-specific product(s) can compete for amplification of the target sequence and impair PCR efficiency. A key component of this kit is AccuStart™ Taq DNA polymerase which contains monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation at 95°C, the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase.

Customers Who Bought This Also Bought