Order Entry
ContactUsLinkComponent
Plasmid Maxiprep Kit
E.Z.N.A.® Plasmid Maxi Kit, Omega Bio-tek®
Catalog #: CA101319-396
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Plasmid Maxi Kit, Omega Bio-tek®
Catalog #: CA101319-396
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Maxi
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Maxi
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Plasmid Maxi Kit
  • Size:
    20 Preps
  • Description:
    Plasmid Maxi Kit
  • Cat. no.:
    CA101319-396
  • Includes:
    Solution I, Solution II, Solution III, Buffer HB, DNA Wash Buffer, Elution Buffer, RNase A, HiBind® DNA Maxi Column I (20), 50 mL Collection Tubes (20)
  • Supplier No.:
    D6922-02

Specifications

About this item

The E.Z.N.A.® Plasmid Maxi Kit is a maxi-scale system designed to deliver high-quality plasmid DNA using the spin column format, without costly accessories

The E.Z.N.A.® Plasmid Maxi Kit is a maxi-scale system designed to deliver high-quality plasmid DNA using the spin column format, without costly accessories

The HiBind® DNA Maxi Column facilitates the binding, washing, and elution steps, thus enabling multiple samples to be simultaneously processed. Although yields vary according to plasmid copy number, E.coli strain, and conditions of growth, 50–200mL of overnight culture in LB medium typically produces 0.5–1mg of high copy number plasmid DNA. Up to 500mL of culture may be processed when working with low copy number plasmid. The product is suitable for automated fluorescent DNA sequencing (typical reads exceed 800bp), restriction enzyme digestion, transfection of mammalian cells, transcription in vitro, and other applications.

Customers Who Bought This Also Bought