Order Entry
ContactUsLinkComponent
BLOOD DNA KIT
E.Z.N.A.® Blood DNA Kit, Omega Bio-tek
Catalog #: CA101319-348
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Blood DNA Kit, Omega Bio-tek
Catalog #: CA101319-348
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    Blood DNA Kit
  • Size:
    200 Preps
  • Description:
    Blood DNA Kit
  • Cat. no.:
    CA101319-348
  • Includes:
    Buffer BL, OB Protease, Proteinase Storage Buffer, Buffer HB, DNA Wash Buffer, Elution Buffer, HiBind® DNA Mini Column, 2mL Collection Tube
  • Supplier No.:
    D3392-02

Specifications

About this item

The E.Z.N.A.® Blood DNA Kit provides rapid total DNA isolation from 1–250 µL of fresh and frozen anticoagulated whole blood

This kit allows for simultaneous processing of single or multiple samples in less than 30 minutes. Phenol/chloroform extractions, and time-consuming steps such as precipitation with isopropanol or ethanol have been eliminated. DNA purified with the E.Z.N.A.® Blood DNA method is ready for applications such as PCR, Southern blotting, or restriction enzyme digestion.

The Kit can also be used for the preparation of genomic DNA from buffy coat, serum, plasma, bone marrow, lymphocytes, platelets, and body fluids.

Customers Who Bought This Also Bought