Order Entry
ContactUsLinkComponent
Plasmid DNA Isolation Kit- E.Z.N.A. Fastfilter Endo-Free Plasmid Maxiprep Kit (25 preps). Isolation of up to 1.5mg endotoxin-free Plasmid DNA in less than 30 minutes with maxi-psin column, Lysate clearance syringe included
E.Z.N.A.® Endo-Free Plasmid Maxi Kit, Omega Bio-tek®
Catalog #: CA101319-342
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Endo-Free Plasmid Maxi Kit, Omega Bio-tek®
Catalog #: CA101319-342
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Maxi
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Endo-Free Plasmid Maxi Kit
  • Size:
    25 Preps
  • Description:
    Endo-Free Plasmid Maxi Kit
  • Cat. no.:
    CA101319-342
  • Includes:
    Solution I, Solution II, Buffer N3, RNase A, ETR Binding Buffer, ETR Wash Buffer, Buffer HB, DNA Wash Buffer, Endo-Free Elution Buffer, HiBind® DNA Maxi Columns, 50mL Collection Tubes, 50mL Lysate Clearance Syringes
  • Supplier No.:
    D6926-03

Specifications

About this item

The Endo-Free Plasmid Maxi Kits provide an efficient method for the large-scale isolation of endotoxin-free plasmid DNA

  • Endotoxin-Free
  • Format Maxi

The Endo-Free Plasmid Maxi Kits provide an efficient method for the large-scale isolation of endotoxin-free plasmid DNA

Following lysis, endotoxins are removed from the cleared lysate with the addition of ETR Solution. Plasmid DNA is then further purified by adsorbing to the HiBind® silica membrane. An easy wash step is followed by the elution of the bound plasmid DNA in elution buffer. Up to 200µg of high-quality plasmid DNA can be purified from 30–50mL of overnight culture (D6915), and up to 1mg from 100–200mL of overnight culture (D6926). Purified DNA samples are ready to be used in downstream applications such as automated fluorescent DNA sequencing, restriction enzyme digestion, transfection of mammalian cells, cloning, and PCR amplification.

Customers Who Bought This Also Bought