Order Entry
ContactUsLinkComponent
MICROELUTE GEL EXTRACTION
MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
Catalog #: CA101319-300
Supplier:  Omega Bio-Tek
CAS Number:  
MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
Catalog #: CA101319-300
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Micro
  • Refine target molecule:
    Total DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    MicroElute® Cycle-Pure & Gel Extraction Kit
  • Description:
    MicroElute® Cycle-Pure & Gel Extraction Kit
  • No. of Reactions:
    50 preps
  • Supplier No.:
    D6294-01

Specifications

About this item

MicroElute® Clean Up system, designed for rapid DNA clean up.

  • Elution volumes – 10-15 µL
  • Generation of highly concentrated DNA
  • 10 µg DNA binding capacity
  • Rapid isolation – less than 15 minutes

The E.Z.N.A.® family of products is an innovative system that radically simplifies the extraction and purification of nucleic acids from a variety of sources. The key to this system is the HiBind® matrix that specifically, but reversibly, binds DNA or RNA under optimized conditions allowing proteins and other contaminants to be removed. Nucleic acids are easily eluted with deionized water or a low salt buffer.

The MicroElute® Clean Up system, designed for rapid DNA clean up, includes:

• MicroElute® Cycle-Pure – for direct purification of double or single stranded PCR products (100 bp – 10 kb) from amplification reactions
• MicroElute® Gel Extraction – for extraction of DNA fragments from standard, or low-melt agarose gels in TAE (Tris-acetate/EDTA) or TBE (Tris-borate/EDTA) buffer

Customers Who Bought This Also Bought