Order Entry
ContactUsLinkComponent
PLANT DNA KIT
E.Z.N.A.® Plant & Fungal DNA Kit, Omega Bio-tek®
Catalog #: CA101319-294
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Plant & Fungal DNA Kit, Omega Bio-tek®
Catalog #: CA101319-294
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    E.Z.N.A.® Plant & Fungal DNA Kit
  • Description:
    Plant & Fungal DNA Kit
  • No. of Reactions:
    200 preps
  • Supplier No.:
    D3485-02

Specifications

About this item

Isolate DNA from plant or fungal samples using spin columns.

  • Rapid – DNA isolation under 60 minutes
  • Reliable – Optimized buffer system ensure the reproducible results
  • Safe – No organic extractions
  • High-quality – Purified DNA suitable for most downstream applications

Isolate DNA from plant or fungal samples using spin columns.

The E.Z.N.A.® Plant & Fungal DNA Kit allows for the rapid and reliable isolation of high-quality total cellular DNA from a wide variety of plant or fungal species and tissues. Up to 200 mg wet tissue (or 50 mg dry tissue) can be processed in less than 1 hour. The system combines the reversible nucleic acid-binding properties of Omega Bio-tek’s HiBind® matrix with the speed and versatility of spin column technology to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant and fungal tissue lysates. Purified DNA is suitable for PCR, restriction digestion, and hybridization applications. There are no organic extractions thus reducing plastic waste and hands-on time to allow multiple samples to be processed in parallel.

If using the E.Z.N.A.® Plant & Fungal DNA Kit for the first time, please read this booklet to become familiar with the procedure and its various modifications. Dry or fresh plant or fungal tissue is disrupted and lysed in a specially formulated buffer containing detergent. Proteins, polysaccharides, and cellular debris are subsequently precipitated. Contaminants are further removed by an isopropanol precipitation step. Binding conditions are adjusted and the sample is transferred to a HiBind® DNA Mini Column. Two rapid wash steps remove trace contaminants such as residual polysaccharides and pure DNA is eluted in water or low ionic strength buffer.

Customers Who Bought This Also Bought