Order Entry
ContactUsLinkComponent
Total RNA Kit
E.Z.N.A.® Total RNA Kit I, Omega Bio-tek®
Catalog #: CA101319-240
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Total RNA Kit I, Omega Bio-tek®
Catalog #: CA101319-240
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Total RNA Kit
  • Size:
    50 Preps
  • Description:
    Total RNA Kit
  • Includes:
    TRK Lysis Buffer, RNA Wash Buffer I, RNA Wash Buffer II, DEPC Water, HiBind® RNA Mini Columns, 2 ml Collection Tubes
  • Supplier No.:
    R6834-01

Specifications

About this item

The E.Z.N.A.® Total RNA Kit I provides a simple and rapid method for the isolation of up to 100 µg of total RNA from cultured eukaryotic cells and soft tissues

This kit enables simultaneous multiple sample processing in less than 20 minutes. Normally up to 1x10^7 eukaryotic cells or 30 mg tissue can be used in a single experiment. Purified RNA can be used in many downstream applications such as RT-PCR, Northern Blotting, Nuclease Protection Assay, and in vitro translation.

Customers Who Bought This Also Bought