Order Entry
ContactUsLinkComponent
Tissue Dna Kit 200 Preps
E.Z.N.A.® Tissue DNA Extraction Systems, Omega Bio-tek
Catalog #: CA101319-018
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Tissue DNA Extraction Systems, Omega Bio-tek
Catalog #: CA101319-018
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Silica spin column
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Bacteria
    Blood
    Cultured cells
    FFPE tissues
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Tissue DNA Kit
  • Size:
    200 Preps
  • Sample size:
    30 mg or 5 x 10⁶ Cells
  • Description:
    Tissue DNA Kit
  • Supplier No.:
    D3396-02

Specifications

About this item

Omega Bio-tek® provides multiple options for genomic, viral, and mitochondrial DNA extraction from cultured cells, buccal swabs, mouse tail snips, whole blood, buffy coat, and a variety of animal tissue

The SQ Tissue DNA Kit is a unique scalable system that permits the isolation of high molecular weight DNA from variable starting sample amounts. The tissue is lysed with a detergent-based buffer and proteins are precipitated out with salt solution. The released DNA is purified and concentrated via alcohol precipitation.

The Mag-Bind® Tissue DNA systems are used for preparation of genomic, viral, and mitochondrial DNA from animal tissue, buccal swabs, mouse tails, whole blood, buffy coat, and cultured cells using paramagnetic beads. Magnetic beads can be used in 24, 96, or 384-well formats to fit your sample size, with the flexibility to process manually or with automation. By eliminating retention volume commonly seen with filter-based applications, magnetic beads allow for higher recovery and optimal yield. Magnetic stands can be added to liquid handlers which utilize the SBS format and can be programmed for your application allowing simultaneous processing of multiple plates. The technology allows for higher purity and flexible elution volumes. The Mag-Bind® Tissue DNA Kits are compatible with open formatted instruments such as Hamilton Nimbus, Starlet, Star Plus, Tecan Freedom Evo®, Beckman Coulter Biomek® 2000, 3000, NX, FX, and Thermo Kingfisher® Flex and Kingfisher® Duo.

The E.Z.N.A.® Tissue DNA Kit uses silica membrane technology accompanied by a specialized buffer system to purify DNA from 30mg of tissue or 5x10⁶ cells. The E-Z 96 silica plates can be used with a vacuum manifold, centrifugation, or automated vacuum systems. It is scalable depending on the size of the sample. Up to 10mg of tissue or 2.5 x 10⁶ cells can be purified using the MicroElute® Tissue DNA Kit elution volume.

Customers Who Bought This Also Bought