Order Entry
ContactUsLinkComponent
MOLLUSC DNA KIT
E.Z.N.A.® Mollusc & Insect DNA Kit, Omega Bio-Tek
Catalog #: CA101319-002
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Mollusc & Insect DNA Kit, Omega Bio-Tek
Catalog #: CA101319-002
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Invertebrates
  • Target molecule:
    DNA
  • Description:
    E.Z.N.A.® Mollusc & Insect DNA Kit
  • Description:
    Mollusc & Insect DNA Kit
  • No. of Reactions:
    5 preps
  • Supplier No.:
    D3373-00

Specifications

About this item

Isolate DNA from molluscs and insects using spin columns.

  • DNA isolation under 20 minutes following lysis
  • Optimised buffer system guarantee pure DNA
  • Purified DNA suitable for any application
  • Isolate DNA from a wide range of samples

The E.Z.N.A.® Mollusc & Insect DNA Kit is designed for efficient recovery of genomic DNA from molluscs, insects, arthropods, roundworms, flatworms, and other invertebrate tissue samples rich in mucopolysaccharides. This kit can also be used with formalin preserved material or invertebrates frozen or preserved in alcohol or DNE solution.

The procedure relies on the well established properties of the cationic detergent, cetyltrimethyl ammonium bromide (CTAB), in conjunction with the selective DNA binding technology of Omega Bio-tek’s HiBind® matrix. Samples are homogenized and lysed in a high salt buffer containing CTAB and extracted with chloroform to remove mucopolysaccharides. Following a rapid alcohol precipitation step, DNA is bound to the HiBind® DNA Mini Column upon adjustment of binding conditions and further purified through multiple wash steps. In this way, salts, proteins and other contaminants are removed to yield high-quality genomic DNA suitable for downstream applications such as endonuclease digestion, thermal cycle amplification, and hybridization techniques.

Customers Who Bought This Also Bought