Order Entry
ContactUsLinkComponent
Plasmid Miniprep Kit I
E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Catalog #: CA101318-896
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Catalog #: CA101318-896
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Midi spin
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Plasmid Mini Kit, Q-Spin
  • Size:
    200 Preps
  • Description:
    Plasmid Mini Kit, Q-Spin
  • Includes:
    Solution I, Solution II, Solution III, Buffer HB, DNA Wash Buffer, Elution Buffer, RNase A, HiBind® DNA Column I (200), 2 mL Collection Tubes (200)
  • Supplier No.:
    D6942-02

Specifications

About this item

The E.Z.N.A.® Plasmid Mini Kit I is designed to isolate up to 30µg of high-quality plasmid DNA from 1–5mL bacterial cultures in 30 minutes or less

The E.Z.N.A.® Plasmid Mini Kit I is designed to isolate up to 30µg of high-quality plasmid DNA from 1–5mL bacterial cultures in 30 minutes or less

Plasmid DNA purification is simplified with HiBind® Mini Column technology into three quick steps: Bind, Wash, and Elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, and manual/automated fluorescent DNA sequencing. E.Z.N.A.® Plasmid Mini Kit I is available in V- Spin and Q-Spin column formats.

Certifications: UL Listed.
Customers Who Bought This Also Bought