Order Entry
ContactUsLinkComponent
CYCLE PURE KIT V SPIN
E.Z.N.A.® Cycle-Pure Kit, Omega Bio-tek
Catalog #: CA101318-890
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Cycle-Pure Kit, Omega Bio-tek
Catalog #: CA101318-890
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Cycle-Pure Kit , V-Spin
  • Size:
    200 Preps
  • Description:
    Cycle-Pure Kit , V-Spin
  • Cat. no.:
    CA101318-890
  • Includes:
    Buffer CP, DNA Wash Buffer, Elution Buffer, Hibind® DNA Mini Column, 2mL Collection Tubes
  • Supplier No.:
    D6492-02

Specifications

About this item

The E.Z.N.A.® Cycle-Pure Kit is designed for the rapid purification of single- or double-stranded DNA from PCR or other enzymatic reactions

The convenient spin-column format eliminates the need for expensive resins or toxic organic compounds such as phenol and chloroform, thereby making it possible to process multiple samples in parallel. Purified DNA can be used in T-A ligations, sequencing, restriction enzyme digestion, and various other labeling reactions.

The purification procedure completely removes primers, nucleotides, enzymes, salts, and other impurities from DNA samples. The DNA sample is simply mixed with CP buffer and spun through the HiBind® DNA column. The DNA bound to the HiBind® Matrix is washed and the clean concentrated DNA is eluted with deionized water or Elution Buffer.

Customers Who Bought This Also Bought