Order Entry
ContactUsLinkComponent
Optix* Dispensers
Optifix® Dispensers, MilliporeSigma
Catalog #: CA10108044-1
Supplier:  MilliporeSigma
Optix* Dispensers
Optifix® Dispensers, MilliporeSigma
Catalog #: CA10108044-1
Supplier:  MilliporeSigma
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Graduation Interval:
    0.5 ml
  • Volume:
    6 - 30 ml
  • Cat. no.:
    CA10108044-1
  • Increments:
    0.5 ml
  • Supplier No.:
    10108044-1

Specifications

About this item

Optifix® Dispensers are available in four models, each of which is designed to increase efficiency and provide additional safety to daily laboratory operations. These dispensers are manufactured to the highest quality standards, ensuring accurate, reliable, and easy dispensing.

The OPTIFIX® SOLVENT dispenser with vapor trap is designed specifically to trap organic vapors, moisture, and/or gases before they enter the bottle and contaminate your solvent. The solid PTFE vapor trap may be filled with an absorbent of choice: charcoal, molecular sieve, silica gel, or t.h.e.® desiccant and easily refilled as needed.

Optifix® Dispensers are available in four models, each of which is designed to increase efficiency and provide additional safety to daily laboratory operations. These dispensers are manufactured to the highest quality standards, ensuring accurate, reliable, and easy dispensing.

Ideal for critical dispensing applications, the Optifix® BASIC dispenser is used for aqueous solutions, light acids, and crystallizing substances

Ideal for critical dispensing applications, the Optifix® BASIC dispenser is used for aqueous solutions, light acids, and crystallizing substances

The Optifix® SOLVENT dispenser is useful for all organic solvents and materials that swell PTFE. The piston is glass. The Optifix® SOLVENT dispenser with vapor trap is designed specifically to trap organic vapors, moisture, and/or gases before they enter the bottle and contaminate the solvent.

The Optifix® SAFETY dispenser is for strong fuming acids, brominated compounds, and strong-smelling reagents. The piston is glass sheathed in PTFE and made with an additional lipseal to prevent fumes from escaping.

The Optifix® HF dispenser is for hydrofluoric acid. The dispenser construction is almost all PTFE.

The piston is glass sheathed in thick PTFE.

Ordering information: Complete with intake tube and Certificate of Precision. Bottles are not included. BASIC and SOLVENT models also include a set of three polypropylene adapters (38mm, 40mm, and 45mm). For the SAFETY and HF models, a PTFE adapter is recommended (see EM-109050-1 series).
Customers Who Bought This Also Bought