To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Terrific broth complete with animal-free soytone
- Sterility:Sterile
- Cat. no.:CA100219-868
- Volume:1 L
- Supplier No.:T7660
Specifications
About this item
For the growth of recombinant E.coli
- No animal-based components
- Sterile
Due to the enriched formulation of TB, strains have an extended growth phase and this results in high cell yields. TB is commonly used for plasmid purification and protein production. Ingredients: 2% animal-free soytone (20 g/L), 2.4% yeast xxtract (24.0 g/L), 0.4% glycerol (4.0 g/L), 72 mM dipotassium phosphate, 17 mM mono potassium phosphate. Ambient shipping conditions. Store at room temperature.