ContactUsLinkComponent
ORION ROSS SURE-FLOW EPOXY PH
Orion™ ROSS® Sure-Flow® Epoxy Combination pH Electrode, Thermo Scientific
Catalog #: CA10010-770
ORION ROSS SURE-FLOW EPOXY PH
Orion™ ROSS® Sure-Flow® Epoxy Combination pH Electrode, Thermo Scientific
Catalog #: CA10010-770
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    120 mm
  • Temperature range:
    0...100 °C
  • Cat. no.:
    CA10010-770
  • pH Range:
    0 - 14
  • Body Material:
    Epoxy
  • Internal Reference:
    ROSS
  • Junction:
    Sure-Flow
  • Connector:
    BNC
  • Supplier No.:
    8165BNWP
Specifications
About this item
Allows for field (on-site) soil pH measurement where stability and high performance are desired
Features the unique liquid-to-liquid Sure-Flow® junction, rugged and durable construction, and a removable protective sleeve/bulb guard.
Customers Who Bought This Also Bought