Specifications
- Clothing Size:One size
- Garment application:Food,Industrial,Chemical protection,Pharmaceutical
- Length:not available
- Sleeve or leg length:Long
- Color:Yellow
- Cat. no.:CA10010-264
- Chest Size:not available
- Supplier No.:QC278BYL0000
- Sleeve:not available
Specifications
About this item
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
- Neck to knee coverage
- Elastic wrists
- Neck loop with snaps and waist ties
- Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Knee-length apron gown design features attached sleeves for enhanced frontal protection. Tightly sewn seams are covered with garment fabric to reinforce seam and reduce potential for particle penetration. Aprons also feature an open back for improved wearer comfort. Bound seams around neck opening feature snap closures and ties at the waist. Elastic openings at wrists allow for a tighter fit.