ContactUsLinkComponent
Tryptic Soy (CASO) Agar
Tryptic Soy Agar Dehydrated Culture Media, MilliporeSigma
Catalog #: CA1.05458.0500
Supplier:  MilliporeSigma
CAS Number:  
Tryptic Soy (CASO) Agar
Tryptic Soy Agar Dehydrated Culture Media, MilliporeSigma
Catalog #: CA1.05458.0500
Supplier:  MilliporeSigma
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Packaging:
    Plastic bottle
  • Description:
    Tryptic Soy Agar
  • Size:
    500 g
  • Cat. no.:
    CA1.05458.0500
  • Supplier No.:
    1.05458.0500
Specifications
About this item
Media is granulated.
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.
Customers Who Bought This Also Bought