Calcium Gluconate Gel, Safecross® First Aid
Catalog #: CA06645
Supplier: Safecross First Aid Limited
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Calcium Gluconate Gel
- Size:30 g
- Cat. no.:CA06645
- Supplier No.:6645
Specifications
About this item
Calcium gluconate gel is designed to mitigate the effects of hydrofluoric acid on skin.
- Antidote for hydrofluoric acid skin burns
- Heals and relieves pain
- Reduces the risk of hypocalcaemia
- Compact; fits into first aid kits and pockets for fast response in an emergency
Easy and painless to apply; creates no risk of increasing tension in the tissues.
Customers Who Bought This Also Bought