OmniPur® PBS Liquid Concentrate 10X, Calbiochem®, MilliporeSigma
Catalog #: CA-EM6505
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:10X
- Packaging:Cubitainer
- Description:Phosphate Buffered Saline
- Size:4 L
- Supplier No.:6505
Specifications
About this item
Molecular biology grade 10X PBS liquid concentrate.
- Clear, colorless liquid
- DNase- and RNase-free
- Intended for laboratory and manufacturing use only
- Not for drug, food or household use
- Freezable
Each liter of concentrate makes 10L of 1X Phosphate Buffered Saline (137mM Sodium Chloride, 2.7mM Potassium Chloride, 10mM Phosphate Buffer).
Customers Who Bought This Also Bought