Order Entry
ContactUsLinkComponent
TRYPTICASE SOY AGAR PK100
BBL™ Trypticase™ Soy Agar (TSA) Dehydrated and Prepared Culture Media, BD Biosciences
Catalog #: CA-90001-286
Supplier:  BD
CAS Number:  
BBL™ Trypticase™ Soy Agar (TSA) Dehydrated and Prepared Culture Media, BD Biosciences
Catalog #: CA-90001-286
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Trypticase™ Soy Agar, Plated
  • Size:
    100/sp
  • Cat. no.:
    CA-90001-286
  • Supplier No.:
    221283

Specifications

About this item

Trypticase Soy Agar is a general-purpose medium for microbiology which supports the growth of fastidious as well as nonfastidious microorganisms.

  • Contains casein and soy peptones for a source of general growth
  • Sodium chloride maintains osmotic equilibrium

Lecithin and polysorbate 80, two commonly used neutralizers, are reported to inactivate residual disinfectants when the sample is being collected.

Customers Who Bought This Also Bought