RODAC™ Plate, Specialty Petri Dish, Sterile, BD Biosciences
Catalog #: CA-90000-004
Supplier: BD
Specifications
- Description:RODAC® Plate
- Cat. no.:CA-90000-004
- Supplier No.:210340
Specifications
About this item
RODAC® (Replicate Organism Detection and Counting) was developed for environmental studies to determine levels of microbial contamination of surfaces
When plate is filled with 12–13mL of media, the media surface is raised to a convex shape. Area of agar bed is approximately 25cm2 with Quebec-style grids of one centimeter squares. A cake-box type lid protects the agar. Polystyrene, with cover. Packaged 20 per sleeve.
Customers Who Bought This Also Bought