FMOC-O-tert-butyl-L-tyrosine
Catalog #: B-1255.0025BA
Supplier: Bachem Americas
CAS Number: 71989-38-3
Specifications
- Size:25 g
- Cat. no.:B-1255.0025BA
- Supplier No.:B-1255.0025
Specifications
About this item
Synonyms: N-α-Fmoc-o-t-butyl-L-Tyrosine,N-Fmoc-O-tert-butyl-L-tyrosine,Nalpha-Fmoc-O-tert-butyl-L-tyrosine,O-tert-Butyl-Nalpha-Fmoc-L-tyrosine,Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 459.54 g/mol
Storage Temperature:Ambient
EINECS:276-262-7
Customers Who Bought This Also Bought