Rheodyne MXT HPLC automated two position, six port, very high pressure switching valve
Catalog #: 97058-392
Supplier: Rheodyne
Rheodyne MXT HPLC automated two position, six port, very high pressure switching valve
Catalog #: 97058-392
Supplier: Rheodyne
Customers Who Bought This Also Bought
Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®
Supplier: BDH
$720.66 to $3,438.57
Bard-Parker® Protected Disposable Scalpels, Sterile, Aspen Surgical
Supplier: Aspen Surgical Products
$68.14 to $587.35
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
$411.94 to $1,303.38
Dichloromethane ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$389.38 to $1,030.72
Magnesium sulfate ≥98.0%, Reagent Grade, VWR Chemicals BDH®
Supplier: VWR International
$158.98 to $1,637.22