Septa
Catalog #: 97057-250
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Supplier: VWR International
$290.42 to $873.69
Lauryl Sulfate Broth Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
$319.37 to $1,392.57
Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$795.94 to $2,497.03
Nitric acid 69.0 - 70.0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$1,076.52
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) surfactant, Millipore®
Supplier: MilliporeSigma
$329.65 to $1,938.83