Decapper for Vials, MicroSlov
Catalog #: 97054-996
Supplier: MicroSolv Technology Corp
Specifications
- Description:Decapper Tool
- Cat. no.:97054-996
- Seal Size:20 mm
- Supplier No.:95025-DC
Specifications
About this item
These decapper tools are used for detaching aluminum Headspace Caps.
Customers Who Bought This Also Bought