Plastic Vials, Caps and Convenience Kits for LCMS and HPLC
Catalog #: 97052-686
Supplier: MicroSolv Technology Corp
Specifications
- Cap included:No
- Capacity:0.70 ml
- Sterility:Non sterile
- Color:Clear
- Description:Vials, AQ™, LCMS Compatible
- Material:Plastic
- Cat. no.:97052-686
- Diameter ×H:12×32 mm
- Maximum Capacity:0.70 ml
- Bottom Style:Flat bottom
- Supplier No.:9572S-MS
Specifications
About this item
Sample safe clear or amber plastic vials for biological, ionic and sensitive analytes.
- Great alternative to glass in many configurations
- Reduce or eliminate adsorption
These vials have excellent cost to performance value. Plastic specifications include no extractables with standard HPLC solvents and meets or exceeds demanding lab requirements to have minimal impact on sensitive samples. The bioinert surface is excellent for use with biological compounds such as many proteins, peptides, nucleic acids, ion-chromatography and analysis of PFAS.
Convenient kit packs with plastic vials and caps make it easy to manage and keep your sample vials clean. LCMS vials are made from a special virgin PP that is excellent with aqueous LCMS and chosen when testing biological, ionic or other compounds that interact with glass.
Customers Who Bought This Also Bought