Caps for Autosampler Vials, Screw Top, MicroSolv™/Basik™
Catalog #: 97052-570
Supplier: MicroSolv Technology Corp
Specifications
- Cap Size:9-425 mm
- Cap Type:Silicone/PTFE Septa
- Description:Black Polypropylene with white silicone
- Cat. no.:97052-570
- Cap Color:Black
- Supplier No.:9502S-E-15K