Caps, BASIK™, MicroSolv
Catalog #: 97052-468
Supplier: MicroSolv Technology Corp
Specifications
- Cap Size:9-425 mm
- Cap Type:Screw cap
- Septum:Clear Silicone Rubber / PTFE, ultra-pure
- Material:PP
- Cat. no.:97052-468
- Cap Color:Blue
- Supplier No.:9502S-06-LCC