Caps
Catalog #: 97052-392
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$78.90 to $874.91
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
VRB (Violet Red Bile) Agar, Fluorocult®, Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
$1,692.83
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
$99.02 to $485.35