WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
Catalog #: 97050-386
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Septum:PTFE disk
- Description:Snap cap, bulk pail
- Cat. no.:97050-386
- Cap Color:Natural
- Supplier No.:11-0051N-B
Specifications
About this item
These snap-top caps eliminate the need for crimping and decapping tools.
- Manufactured from PE
- Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Customers Who Bought This Also Bought