Disposable Syringes, MicroLiter, DWK Life Sciences
Catalog #: 97048-639
Supplier: DWK Life Sciences (KIMBLE)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Volume:1000 µl (1 ml)
- Description:Tuberculin Disposable Syringe
- Supplier No.:F5-A1
Specifications
About this item
MicroLiter's syringes feature polypropylene housings that are solvent-resistant and clearly marked for media and pore size.
Ordering information: Glass fiber prefilters and syringe filters are available in a variety of media types and pore sizes.
Customers Who Bought This Also Bought