WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Catalog #: 97047-918
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Septum:Natural PTFE/Blue Silicone
- Description:AlumiTin™ Small Opening
- Cat. no.:97047-918
- Cap Color:Green
- Supplier No.:20-0050ATS
Specifications
About this item
MicroLiter's 20 mm caps are available in multiple colors and septa types.
MicroLiter offers products for both LC and GC applications.
Customers Who Bought This Also Bought