WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Catalog #: 97047-866
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Septum:PTFE/butyl
- Description:Septa
- Cat. no.:97047-866
- Supplier No.:20-0030
Specifications
About this item
These septa are used for 20 mm headspace vial crimp seals.
Designed for optimum performance in a variety of applications and heat ranges.
Customers Who Bought This Also Bought