WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
Catalog #: 97047-174
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Septum:PTFE/silicone
- Description:Snap cap
- Cat. no.:97047-174
- Cap Color:Black
- Supplier No.:11-0053K
Specifications
About this item
These snap-top caps eliminate the need for crimping and decapping tools.
- Manufactured from PE
- Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Customers Who Bought This Also Bought