Septa for 10-425 Screw-Thread Caps, MicroLiter
Catalog #: 97046-948
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Septum:PTFE/silicone/PTFE
- Description:Septa, for 10-425 caps
- Thickness:0.040"
- Cat. no.:97046-948
- Supplier No.:10-0040
Specifications
About this item
Septa for the original wide-opening vials from MicroLiter are ideal for chromatography applications where vials remain stationary.
Packaging: Class 10000 Clean Room Packaging includes Dispenser Pack septa for proper storage and handling on the bench.
Customers Who Bought This Also Bought