Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
Catalog #: 97046-220
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Ø:9 mm
- Septum:Tefzel®/Silicone
- Description:Blue Polypropylene
- Supplier No.:09-0022B
Specifications
About this item
MicroLiter's 9 mm screw-thread caps are available in multiple colors and septa types.
Customers Who Bought This Also Bought