Specifications
- Garment application:Laboratory
- Length:44.5 cm (171/2")
- Shoe size:Universal
- Color:Blue
- Description:Anti-skid shoe covers
- Tearing strength:7.9 (Widthwise Yarns)
- Particle shedding (Helmke Drum):Level II
- Bacterial filtration efficiency:0.55
- Weight:36.89 g/m²
Specifications
About this item
These spunbonded polypropylene (SPP) shoe covers protect against dirt, grime, and certain dry particulates in nonhazardous environments
These spunbonded polypropylene (SPP) shoe covers protect against dirt, grime, and certain dry particulates in nonhazardous environments. The fabric is strong, lightweight, and breathable. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications. Shoe covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
- Economical, disposable first-line barrier
- Maximum breathability
- Low linting
Available with rubber-coated soles for secure footing on wet or dry surfaces. Elastic top and bottom. Shoe covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
The fabric is strong, lightweight, and breathable. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.