Order Entry
ContactUsLinkComponent
Model 716 bench clamp
VWR® Talon® Cylinder Bench Clamps
Catalog #: 97035-530
Supplier:  VWR International
Product available on GSA Advantage®
Model 716 bench clamp
VWR® Talon® Cylinder Bench Clamps
Catalog #: 97035-530
Supplier:  VWR International
Product available on GSA Advantage®

Specifications

  • Description:
    Bench Clamp with Safety Message Strap and Chain
  • Cat. no.:
    97035-530

Specifications

About this item

Rugged cast aluminum cylinder clamps secure gas cylinders to benches, tables or other flat surfaces up to 6.4 cm (21/2") thick.

  • Rugged design
  • Cast aluminum construction
  • Non-slip spring catch and buckle
  • Easily adjustable
  • Dimensions: 8.3L x 13.3W x 16.5H cm (31/4 x 51/4 x 61/2")

The 2.5 cm (1") wide, 137.2 cm (54") long nylon strap features a nickel-plated, non-slip spring catch and buckle for easy adjustment around cylinders 10.2 – 35.6 cm (4 – 14") in diameter.

Customers Who Bought This Also Bought