HPLC Columns, YMC Carotenoid
Catalog #: 97030-718
Supplier: YMC America,Inc
Specifications
- Column Format:Analytical
- I.D.:4.60 mm
- Length:250 mm
- Particle size:5 µm
- Endcapped:Yes
- USP code:L62
- pH range:2 - 7.5
- Cat. no.:97030-718
- Packing Material:Silica
- Phase:Carotenoid C30
- Supplier No.:CT99S05-2546WT
Specifications
About this item
The Carotenoid C30 stationary phase provides sufficient phase thickness to enhance interaction with long chain molecules therefore, geometric and positional isomers of conjugated double-bonding systems are recognized and resolved.
- These columns is much more hydrophobic and lipophilic
- Even when pure organic eluents are applied, many sample solutes are retained
- Applicable for LC-MS
Carotenoids are found in a variety of natural sources including fruits and vegetables. In addition, carotenoids are considered as potential drugs for cancer intervention and/or prevention. Despite the complexity of carotenoid extracts and the minor shape differences between carotenoid isomers, the separation, identification, and quantitation of these compounds can be achieved using these columns.
Customers Who Bought This Also Bought