Finnpipette® Bottle Top Dispenser, Thermo Scientific
Catalog #: 97004-414
Supplier: Thermo Fisher Scientific
Specifications
- Accuracy:±180 μl
- Volume:5 - 30 ml
- Precision:60.0 μL
- Supplier No.:4421160
Specifications
About this item
Finnpipette Dispensers protect users even when handling aggressive liquids
- Ergonomically designed
- Non-slip piston handle
- Excellent chemical and thermal resistance
- Precise self-locking system in the volume setting
The dispensers are available in six models, covering volumes between 0.2 and 60mL. There is a universal fit for reagent bottles, and the dispensing head cap prevents dripping.
Customers Who Bought This Also Bought