Order Entry
ContactUsLinkComponent
VWR Universal Platform Stainless Steel 18Inx24In
Accessories for VWR Standard and Advanced Orbital Shakers
Catalog #: 97003-590
Supplier:  VWR International
Product available on GSA Advantage®
Accessories for VWR Standard and Advanced Orbital Shakers
Catalog #: 97003-590
Supplier:  VWR International
Product available on GSA Advantage®

Specifications

  • Description:
    Universal Platform, 45.7×61 cm (18×24")
  • Cat. no.:
    97003-590

Specifications

About this item

Customers Who Bought This Also Bought