Axygen® Single- and Multi-Well Reservoirs, Corning
Catalog #: 95042-900
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Capacity:290 ml
- Sterility:Sterile
- Well Volume:10 ml
- Color:Translucent
- Description:Multi-well reservoir, 8 channel trough, low profile
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:95042-900
- Packaging:Individually wrapped
- Supplier No.:RES-MW8-LP-SI
Specifications
About this item
These polypropylene reagent vessels are appropriate for automated and manual applications.
- Reservoirs are made of clear virgin polypropylene for excellent chemical resistance
- All reservoirs feature an SBS footprint for compatibility with most robotic systems
- Flat-bottom reservoirs are single-well format and are guaranteed flat
These polypropylene reagent vessels are appropriate for automated and manual applications.
Trough reservoirs incorporate flow troughs that hold the sample across the length or breadth of the reservoir in either 8- or 12-trough configurations. Diamond-bottom reservoirs feature 96- and 384-well profiles that are ideal for 8- and 12-channel pipettor applications.
Ordering information: Sterile reservoirs are individually wrapped.
Customers Who Bought This Also Bought