Cytology Spin Slides, Azer Scientific
Catalog #: 95042-274
Supplier: Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Specifications
- Description:Cytospin slide, single well
- Supplier No.:ESCS-096
Specifications
About this item
Cytology Spin slides are compatible with Thermo Shandon centrifuge systems and are designed to aid in the quick and easy location of the centrifuged smear.
- Thermo Shandon centrifuge system-compatible
- Single or double wells available
- Adhesive coating available
- Well placement in the same position of centrifuge specimen deposit
Available with single or double wells.
Slides feature well placement in the exact position where the centrifuge specimen is deposited. Available with or without adhesive coating.
Packaging: 72 slides per box, two boxes per case
Customers Who Bought This Also Bought