Specifications
- Description:TG1 Electrocompetent Cells (≥2 x 10^10 cfu/µg DNA)
- Size:24 reactions
- Genotypes:TG1: [F' traD36 proAB lacIqZ ΔM15] supE thi-1 Δ(lac-proAB) Δ(mcrB-hsdSM)5(rK - mK -)
- Cat. no.:95040-452
- Packaging:DUOs
- Supplier No.:60502-2
Specifications
About this item
Lucigen’s Phage Display Electrocompetent Cells are perfect for antibody phage display or peptide phage display library creation
Lucigen offers the following Phage Display Electrocompetent Cells:
TG1 Electrocompetent Cells: An amber suppressor strain (supE) prepared as highly efficient (≥4 × 10^10 cfu/µg) electrocompetent cells for phage display library screening. These cells are used for phage display and protein expression.
SS320 Electrocompetent Cells: A non-amber suppressor strain (sometimes called MC1061F') prepared as highly efficient (≥4 × 10^10 cfu/µg) electrocompetent cells for phage display library screening. These cells have the highest available transformation efficiency of any strain used for phage display.
ER2738 Electrocompetent Cells: An amber suppressor strain (glnV) prepared as highly efficient (≥2 × 10^10 cfu/µg) electrocompetent cells for phage display library screening. This strain is recommended for use with New England Biolab's Ph.D.™ Phage Display Kits.
These electrocompetent cells are also ideal for creating larger libraries to speed discovery. Lucigen offers the highest efficiency TG1 competent cells available (≥4 × 10^10cfu/µg), and is your only source for electrocompetent SS320 and ER2738 cells. Amber suppressor and non-amber suppressor lines are available for maximum flexibility.