ContactUsLinkComponent
100 BP PLUS DNA LADDER 500UL
DNA Ladders/Molecular Weight Markers, Bioneer
Catalog #: 95027-584
Supplier:  Bioneer
CAS Number:  
DNA Ladders/Molecular Weight Markers, Bioneer
Catalog #: 95027-584
Supplier:  Bioneer
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    100 bp Plus DNA ladder
  • Cat. no.:
    95027-584
  • Volume:
    500 µl
  • Supplier No.:
    D-1035
Specifications
About this item
25/100bp mixed DNA ladder is specially designed for determining the size of double strand DNA from 25–2000 base pairs
The DNA Ladder consists of 17 double strand DNA fragments ranging in size from 25 to 200 bp in 25 bp increments, and additional fragments of 300, 400, 500, 600, 700, 800, 900, 1000, 2000 bp. The 150, 500, 1000, and 2000 bp bands are approximately two to three times brighter for easy identification. 100bp DNA Ladder is specially designed for determining the size of double strand DNA from 100 to 2000 base pairs. The DNA Ladder consists of 13 double strand DNA fragments ranging in sizes from 100 to 1000 bp in 100 bp increments, and additional fragments of 1200, 1600, 2000 bp. The 500, 1000, and 2000 bp bands are two to three times brighter for easy identification. 100 bp plus DNA ladder is specially designed for determining the size of double strand DNA from 100 to 10,200 base pairs. The DNA ladder consists of 19 double strand DNA fragments ranging in sizes from 100 to 1000 bp in 100 bp increments, and additional fragments of 1200, 1600, 2000, 2961, 4000, 5007, 5991, 8000, and 10,200bp. The 500, 1000, and 2000 bp bands are two to three times brighter for easy identification.
1Kb DNA Ladder is specially designed for determining the size of double strand DNA from 500–10,000 base pairs. The DNA ladder consists of DNA fragments 0.5, 1.0, 1.6, 2.0, 3.0 (2.961), 4.0, 5.0 (5.007), 6.0 (5.991), 8.0, and 10.2 (10.200) kilo base pairs. The 2.961Kb band is approximately two to three times brighter for easy identification.
Lambda DNA/Hind III marker is designed for determining double strand DNA from 125 to 23,130 base pairs. The marker is prepared by digesting Lambda DNA to completion with Hind III, followed by phenol inactivation of the enzyme.
Lambda DNA/EcoR I marker is designed for determining double strand DNA from 3530 to 21,226 base pairs. The marker is prepared by digesting Lambda DNA to completion with EcoR I, followed by phenol inactivation of the enzyme.
Lambda DNA / EcoR I + Hind III marker is designed for determining double strand DNA from 125 to 21,226 base pairs. The marker is prepared by digesting Lambda DNA to completion with both EcoR I and Hind III, followed by phenol inactivation of the enzyme.
Ordering information: The DNA ladder can be applied directly onto an agarose gel. Marker 95030-432 is not designed for quantitation. Markers 95027-592, -596, and -600 may form additional bands due to annealing of cohesive end, and these fragments can be separated by heating at 60–65°C for 5 minutes and then cooling on ice for three minutes.
Caution: For research use only. Not for use in diagnostic or therapeutic procedures. Do not heat before loading. Repeated freezing and thawing should be avoided.
Customers Who Bought This Also Bought