Order Entry
ContactUsLinkComponent
Nitrate Test Kit
Nitrate, CHEMets Visual Kit, CHEMetrics
Catalog #: 95026-866
Supplier:  CHEMetrics
Small Business Enterprise
CAS Number:  
Nitrate Test Kit
Nitrate, CHEMets Visual Kit, CHEMetrics
Catalog #: 95026-866
Supplier:  CHEMetrics
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Cadmium reduction
  • Description:
    Nitrate CHEMets Visual Test Kit
  • Cat. no.:
    95026-866
  • Range:
    0 - 225 ppm as N
  • Supplier No.:
    K-6909A

Specifications

About this item

Nitrate is reduced to nitrite using cadmium as the reducing agent. The resulting nitrite concentration is then determined colorimetrically. This method is applicable to drinking and surface waters, as well as domestic and industrial wastes.

  • Ideal for low- to medium-range colorimetric analysis of water quality

CHEMets® test kits utilize self-filling ampules and vacuum pulls the sample up from a snap cup into the ampule when the tip is broken off. After tilting the ampule several times to mix the sample and reagent and waiting for the color to develop (usually 2 minutes or less), the analyst quantifies the result using the provided cylindrical comparator (for concentrations ≤1ppm) or flat comparator (for concentrations ≥1ppm).

Nitrite will interfere with this test. Results are expressed as ppm (mg/L) NO₃-N or NO₃.

Nitrate is the most completely oxidized form of nitrogen. It is formed during the final stages of biological decomposition, either in wastewater treatment facilities or in natural water supplies. Low-level nitrate concentrations may be present in natural waters. However, a Maximum Contaminant Level of 10 ppm nitrate-nitrogen has been established for drinking water by the USEPA.

Delivery: Kits come in a plastic case and contain everything needed to perform 30 tests (except distilled water): Refill, Comparator, 25 mL sample cup, syringe, reaction tube with lid, and instructions.
Customers Who Bought This Also Bought