DOT Labels, National Marker
Catalog #: 94012-150
Supplier: National Marker
Specifications
- Description:DOT labels
- Cat. no.:94012-150
- Legend:Flammable 3
- Dimensions:101.6×101.6 mm (4×4")
- Supplier No.:DL158AP
Specifications
About this item
Apply proper labels onto the outside of packing containers indentifying the appropriate hazard.
Certifications: Meets DOT 49 CFR 172.519 requirements.
Customers Who Bought This Also Bought